missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine hydrochloride, MedChemExpress

Product Code. 16459126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459126 Supplier Medchem Express Supplier No. HYN0682
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pyridoxine hydrochloride (Pyridoxol; Vitamin B6) is a pyridine derivative. Pyridoxine (Pyridoxol; Vitamin B6) exerts antioxidant effects in cell model of Alzheimer's disease via the Nrf-2/HO-1 pathway.

Pyridoxine hydrochloride (Pyridoxol

Specifications

Chemical Name or Material Pyridoxine hydrochloride
CAS 58-56-0
Color White
Purity Grade Notes Research
Molecular Formula C8H12ClNO3
Quantity 100 mg
Synonym Pyridoxol hydrochlorideVitamin B6 hydrochloride
Solubility Information H2O : ≥ 50 mg/mL (243.14 mM) ∣DMSO : ≥ 50 mg/mL (243.14 mM)
SMILES OCC1=C(CO)C(O)=C(C)N=C1.Cl
Molecular Weight (g/mol) 205.64
Formula Weight 205.64
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.