missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine Hydrochloride, Fisher BioReagents

Product Code. 10256204
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Amber Glass
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10256204 100 g Amber Glass 100g
10758325 10 g Amber Glass 10g
11449292 50 g Amber Glass 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10256204 Supplier Fisher BioReagents Supplier No. BP2677100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Pyridoxine HCl is used as a vitamin (enzyme cofactor).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H12ClNO3
Molecular Weight (g/mol) 205.638
InChI Key ZUFQODAHGAHPFQ-UHFFFAOYSA-N
Synonym pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
PubChem CID 6019
ChEBI CHEBI:30961
IUPAC Name 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
SMILES CC1=NC=C(C(=C1O)CO)CO.Cl

Specifications

Melting Point 214°C
Color White
pH 2.4 to 3.0
Ignition Residue 0.1% max.
Loss on Drying 0.5% max.
Infrared Spectrum Conforms to reference
Assay Percent Range ≥98 %
Packaging Amber Glass
Quantity 100 g
Merck Index 15, 8095
Formula Weight 205.64
Percent Purity 98% min.
Physical Form Powder or Solid
Chemical Name or Material Pyridoxine Hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Pyridoxine Hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.