missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxin hydrochloride (Vitamin B6 hydrochloride), Dr. Ehrenstorfer

Product Code. 30315608
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315608 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315608 Supplier Dr. Ehrenstorfer Supplier No. DREC16652000100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyridoxine Hydrochloride
CAS 58-56-0
Analyte or Component Names Pyridoxine Hydrochloride
Shipping Condition +20°C
Molecular Formula C8 H11 N O3 . Cl H
Certifications/Compliance ISO 17034
Synonym Pyridoxine Hydrochloride, 5-Hydroxy-6-methyl-3,4-pyridinedimethanol hydrochloride, Pyridoxol hydrochloride, 2-Methyl-3-hydroxy-4,5-bis(hydroxymethyl)pyridine hydrochloride, 2-Methyl-3-hydroxy-4,5-di(hydroxymethyl)pyridine hydrochloride, 3-Hydroxy-4,5-bis(hydroxymethyl)-2-methylpyridine hydrochloride, 4,5-Bis(hydroxymethyl)-2-methylpyridin-3-ol hydrochloride, 4,5-Bis(hydroxymethyl)-3-hydroxy-2-methylpyridine hydrochloride, 4,5-Di(hydroxymethyl)-3-hydroxy-2-methylpyridine hydrochloride, 4,5-Dimethylol-3-hydroxy-2-methylpyridine hydrochloride, 5-Hydroxy-6-methyl-3,4-pyridinedicarbinol hydrochloride, Aderomine hydrochloride, Aderoxin, Aderoxine, Becilan, Beesix, Benadon, Bonasanit, Campoviton 6, Hexabetalin, Hexabione hydrochloride, Hexavibex, Hexermin, Hexobion, Pyridipca, Pyridox, Pyridoxin hydrochloride, Pyridoxine chloride, Pyridoxine hydrochloride, Pyridoxine hydrogen chloride, Pyridoxine monohydrochloride, Pyridoxinium chloride, Vitamin B6 hydrochloride, B6 HCl, 3-Hydroxy-4,5-dimethylol-alpha-picoline hydrochloride
InChI Formula InChI=1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H
SMILES Cl.Cc1ncc(CO)c(CO)c1O
IUPAC Name 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
Molecular Weight (g/mol) 205.64
Quantity 100 mg
Formula Weight 205.0506 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.