missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxamine Dihydrochloride, TRC

Product Code. 30135886
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135886 5 g 5g
17960335 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135886 Supplier Toronto Research Chemicals Supplier No. TRCP9917255G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524-36-7
Molecular Formula C8 H12 N2 O2 . 2 Cl H
Molecular Weight (g/mol) 241.12
Synonym 3-Pyridinemethanol, 4-(aminomethyl)-5-hydroxy-6-methyl-, dihydrochloride (9CI), Pyridoxamine, dihydrochloride (8CI), 2-Methyl-3-hydroxy-4-aminomethyl-5-hydroxymethylpyridine dihydrochloride, 4-(Aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol dihydrochloride, 4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride, Pyridorin, Pyridoxamine dichlorohydrate, Pyridoxylamine dihydrochloride
IUPAC Name 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol;dihydrochloride
SMILES Cl.Cl.Cc1ncc(CO)c(CN)c1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H12N2O2.2ClH/c1-5-8(12)7(2-9)6(4-11)3-10-5;;/h3,11-12H,2,4,9H2,1H3;2*1H
Formula Weight 240.0432
Chemical Name or Material Pyridoxamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.