missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal phosphate, MedChemExpress

Product Code. p-200099376
Encompass_Preferred
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 16438806

Brand: Medchem Express HYB1744

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyridoxal phosphate is the active form of vitamin B6, acts as an inhibitor of reverse transcriptases, and is used for the treatment of tardive dyskinesia.

Pyridoxal phosphate is the active form of vitamin B6, acts as an inhibitor of reverse transcriptases, and is used for the treatment of tardive dyskinesia.

Specifications

Chemical Name or Material Pyridoxal phosphate
CAS 54-47-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C8H10NO6P
Quantity 100 mg
Synonym Pyridoxal 5′-phosphate Pyridoxyl phosphate
Solubility Information H2O : 5 mg/mL (20.23 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (4.05 mM; Need ultrasonic)
SMILES O=CC1=C(O)C(C)=NC=C1COP(O)(O)=O
Molecular Weight (g/mol) 247.14
Formula Weight 247.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.