missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal phosphate, MedChemExpress

Product Code. 16438806
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438806 100 mg 100mg
16412359 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438806 Supplier Medchem Express Supplier No. HYB1744
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pyridoxal phosphate is the active form of vitamin B6, acts as an inhibitor of reverse transcriptases, and is used for the treatment of tardive dyskinesia.

Pyridoxal phosphate is the active form of vitamin B6, acts as an inhibitor of reverse transcriptases, and is used for the treatment of tardive dyskinesia.

Spécification

Chemical Name or Material Pyridoxal phosphate
CAS 54-47-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C8H10NO6P
Quantity 100 mg
Synonym Pyridoxal 5′-phosphate Pyridoxyl phosphate
Solubility Information H2O : 5 mg/mL (20.23 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (4.05 mM; Need ultrasonic)
SMILES O=CC1=C(O)C(C)=NC=C1COP(O)(O)=O
Molecular Weight (g/mol) 247.14
Formula Weight 247.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Afficher plus Afficher moins

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.