missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxal Kinase/PDXK Antibody (OTI5H5), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73754MFV450
This item is not returnable.
View return policy
Description
Pyridoxal Kinase/PDXK Monoclonal antibody specifically detects Pyridoxal Kinase/PDXK in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| Pyridoxal Kinase/PDXK | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI5H5 | |
| Flow Cytometry | |
| C21orf124, C21orf97, chromosome 21 open reading frame 124, DKFZp566A071, EC 2.7.1.35, FLJ21324, FLJ37311, MGC15873, MGC31754, MGC52346, PKHchromosome 21 open reading frame 97, PNKhuman pyridoxal kinase, EC 2.7.1.3510FLJ31940, PRED79, pyridoxal (pyridoxine, vitamin B6) kinase, pyridoxal kinase, pyridoxamine kinase, Pyridoxine kinase, vitamin B6 kinase | |
| Full length human recombinant protein of human PDXK(NP_03672) produced in HEK293 cell. | |
| 0.1 mL | |
| metabolism | |
| 8566 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction