missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxal Kinase/PDXK Antibody (OTI5H5), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73754JF646
This item is not returnable.
View return policy
Description
Pyridoxal Kinase/PDXK Monoclonal specifically detects Pyridoxal Kinase/PDXK in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Pyridoxal Kinase/PDXK | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDXK | |
| Full length recombinant protein of human Pyridoxal Kinase/PDXK (NP_03672) produced in HEK293 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI5H5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| C21orf124, C21orf97, chromosome 21 open reading frame 124, DKFZp566A071, EC 2.7.1.35, FLJ21324, FLJ37311, MGC15873, MGC31754, MGC52346, PKHchromosome 21 open reading frame 97, PNKhuman pyridoxal kinase, EC 2.7.1.3510FLJ31940, PRED79, pyridoxal (pyridoxine, vitamin B6) kinase, pyridoxal kinase, pyridoxamine kinase, Pyridoxine kinase, vitamin B6 kinase | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 8566.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction