missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal isonicotinoyl hydrazone, MedChemExpress

Product Code. 16441107
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16441107

Brand: Medchem Express HY114758

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyridoxal isonicotinoyl hydrazone (PIH) is a lipophilic, tridentate Fe-chelating agent that shows high Fe chelation efficacy.

Pyridoxal isonicotinoyl hydrazone (PIH) is a lipophilic, tridentate Fe-chelating agent that shows high Fe chelation efficacy.

Specifications

Chemical Name or Material Pyridoxal isonicotinoyl hydrazone
CAS 737-86-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H14N4O3
Quantity 50 mg
Synonym PIH
Solubility Information DMSO : 14.71 mg/mL (51.38 mM; Need ultrasonic)
SMILES CC1=NC=C(C(/C=N/NC(C2=CC=NC=C2)=O)=C1O)CO
Molecular Weight (g/mol) 286.29
Formula Weight 286.29
Percent Purity 98.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.