missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal Hydrochloride, TRC

Product Code. 30010216
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010216 25 g 25g
30005803 5 g 5g
17961595 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010216 Supplier Toronto Research Chemicals Supplier No. TRCP99172025G
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-22-5
Molecular Formula C8 H9 N O3 . Cl H
Molecular Weight (g/mol) 203.62
Synonym 4-Pyridinecarboxaldehyde, 3-hydroxy-5-(hydroxymethyl)-2-methyl-, hydrochloride (9CI), Pyridoxal, hydrochloride (7CI,8CI), 2-Methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, 3-Hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-Hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride
IUPAC Name 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
SMILES Cl.Cc1ncc(CO)c(C=O)c1O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H9NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,4,10,12H,3H2,1H3;1H
Formula Weight 203.0349
Chemical Name or Material Pyridoxal Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.