missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal Hydrochloride, Fisher BioReagents

Product Code. 10194814
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10194814 1 g Amber Glass 1g
10776785 5 g Amber Glass 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10194814 Supplier Fisher BioReagents Supplier No. BP26751
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Pyridoxal Hydrochloride is useful for labeling amino acids and for their detection in the picomole range.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-22-5
Molecular Formula C8H10ClNO3
Molecular Weight (g/mol) 203.622
InChI Key FCHXJFJNDJXENQ-UHFFFAOYSA-N
Synonym pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
PubChem CID 6171
IUPAC Name 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
SMILES CC1=NC=C(C(=C1O)C=O)CO.Cl

Specifications

Melting Point 171°C
Color White
pH 2.1
Packaging Amber Glass
Quantity 1 g
Merck Index 15, 8091
Formula Weight 203.63
Physical Form Powder or Solid
Chemical Name or Material Pyridoxal Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.