missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal 5-Phosphate (Monohydrate), Fisher BioReagents

Product Code. 10033284
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10033284 25 g Amber Glass 25g
10516204 1 g Amber Glass 1g
10516394 5 g Amber Glass 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10033284 Supplier Fisher BioReagents Supplier No. BP267625
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • PLP is useful for modifying lysyl and valyl residues and functions as a coenzyme in transamination reactions by forming a Schiff-base linkage with lysine groups on aminotransferase
  • PLP is an active form of vitamin B6
  • Also serves as a coenzyme in some decarboxylation and deamination reactions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-47-7
Molecular Formula C8H10NO6P
Molecular Weight (g/mol) 247.14
MDL Number MFCD00006333
InChI Key NGVDGCNFYWLIFO-UHFFFAOYSA-N
Synonym pyridoxal phosphate, codecarboxylase, pyridoxal 5-phosphate, pyridoxal 5'-phosphate, pyridoxal p, biosechs, hairoxal, pyromijin, vitazechs, hiadelon
PubChem CID 1051
ChEBI CHEBI:18405
IUPAC Name (4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate
SMILES CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O

Specifications

Melting Point 140°C
Color White
Infrared Spectrum Conforms to reference
Assay Percent Range ≥98%
Packaging Amber Glass
Quantity 25 g
Merck Index 15, 8092
Formula Weight 247.14
Percent Purity ≥98%
Physical Form Powder or Solid
Chemical Name or Material Pyridoxal 5-Phosphate, Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.