missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal-5-phosphate hydrate, 98%

Code produit 11459433
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Conditionnement:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
11459433 5 g 5g
11449433 1 g 1g
11469433 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11459433 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur A12323.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 41468-25-1 | C8H10NO6P | 247.14 g/mol

Active form of Vitamin B-6Pyridoxal-5’-phosphate monohydrate is used to modify the lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions and as an inhibitor for purinergic receptors and intracellular. It is involved in decarboxylation and deamination reactions viz. the conversion of dopa into dopamine of glutamate into GABA and histidine to histamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Active form of Vitamin B-6Pyridoxal-5’-phosphate monohydrate is used to modify the lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions and as an inhibitor for purinergic receptors and intracellular. It is involved in decarboxylation and deamination reactions viz. the conversion of dopa into dopamine of glutamate into GABA and histidine to histamine.

Solubility
Soluble in alkaline solutions. Sparingly soluble in water.

Notes
Light sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 41468-25-1
Molecular Formula C8H10NO6P
Molecular Weight (g/mol) 247.14
MDL Number MFCD00006333
InChI Key NGVDGCNFYWLIFO-UHFFFAOYSA-N
Synonym 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate
PubChem CID 38882
IUPAC Name (4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate;hydrate
SMILES CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O

Spécification

Melting Point 142°C
Quantity 5 g
Sensitivity Light sensitive
Merck Index 14,7979
Solubility Information Soluble in alkaline solutions. Sparingly soluble in water.
Formula Weight 265.17 (247.15 Anhydrous)
Percent Purity 98%
Chemical Name or Material Pyridoxal-5-phosphate hydrate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.