missing translation for 'onlineSavingsMsg'
Learn More

Pyridostigmine bromide, MedChemExpress

Product Code. 16490818
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490818 5 g 5g
16422089 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490818 Supplier Medchem Express Supplier No. HYB0207A
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pyridostigmine bromide is a parasympathomimetic and a reversible cholinesterase inhibitor.

Pyridostigmine bromide is a parasympathomimetic and a reversible cholinesterase inhibitor.

Specifications

Chemical Name or Material Pyridostigmine bromide
Physical Form Solid
CAS 101-26-8
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C9H13BrN2O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 50 mg/mL (191.48 mM; Need ultrasonic) ∣H2O : 8.8 mg/mL (33.70 mM; Need ultrasonic and warming)
SMILES C[N+]1=CC=CC(OC(N(C)C)=O)=C1.[Br-]
Molecular Weight (g/mol) 261.12
Formula Weight 261.12
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.