missing translation for 'onlineSavingsMsg'
Learn More

Pyridostigmine bromide, 98%

Product Code. 15618725 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15618725 1 g 1g
15628725 5 g 5g
15638725 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15618725 Supplier Thermo Scientific Acros Supplier No. 461190010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101-26-8 | C9H13BrN2O2 | 261.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-26-8
Molecular Formula C9H13BrN2O2
Molecular Weight (g/mol) 261.12
MDL Number MFCD00079283
InChI Key VNYBTNPBYXSMOO-UHFFFAOYSA-M
Synonym pyridostigmine bromide, mestinon, regonol, kalimin, kalymin, regonal, 3-dimethylcarbamoyl oxy-1-methylpyridin-1-ium bromide, mestinon bromide, 3-dimethylcarbamoyloxy-1-methylpyridinium bromide
PubChem CID 7550
IUPAC Name (1-methylpyridin-1-ium-3-yl) N,N-dimethylcarbamate;bromide
SMILES [Br-].CN(C)C(=O)OC1=CC=C[N+](C)=C1

Specifications

Melting Point 154.0°C
Loss on Drying 2% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Merck Index 15,809
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, chloroform, insoluble in ether, benzene, acetone
Formula Weight 261.12
Percent Purity 98%
Chemical Name or Material Pyridostigmine bromide
Show More Show Less
compliance-icons
Product Identifier
  • Pyridostigmine bromide, 98%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Pyridostigmine bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.