missing translation for 'onlineSavingsMsg'
Learn More

Pyridone 6, MedChemExpress

Product Code. 16452207
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452207 50 mg 50mg
16466068 10 mg 10mg
16460216 2 mg 2mg
16428677 5 mg 5mg
16456466 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16452207 Supplier Medchem Express Supplier No. HY14435

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyridone 6 is a pan-JAK inhibitor, which potently inhibits the JAK kinase family, with IC50s of 1 nM for JAK2 and TYK2, 5 nM for JAK3, and 15 nM for JAK1, while displaying significantly weaker affinities (130 nM to >10 mM) for other protein tyrosine kinases.

Pyridone 6 is a pan-JAK inhibitor, which potently inhibits the JAK kinase family, with IC50s of 1 nM for JAK2 and TYK2, 5 nM for JAK3, and 15 nM for JAK1, while displaying significantly weaker affinities (130 nM to >10 mM) for other protein tyrosine kinases.

Specifications

Chemical Name or Material Pyridone 6
CAS 457081-03-7
Color Brown
Purity Grade Notes Research
Molecular Formula C18H16FN3O
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (323.27 mM)
SMILES FC1=CC2=C(C(NC=C3)=O)C3=C(N=C(C(C)(C)C)N4)C4=C2C=C1
Molecular Weight (g/mol) 309.34
Formula Weight 309.34
Percent Purity 98.84%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.