missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Pyridone 6

Description
Pyridone 6 is a potent pan-JAK inhibitor; ATP-competitive inhibitor of JAK 1/2/3 and Tyk2 (IC50 values are 1, 5, 15 and 1 nM, respectively). Inhibits other kinases tested at 130 nM to >10 μM. Inhibits IL-2- and IL-4-dependent proliferation of CTLL cells and blocks STAT5 phosphorylation. Also inhibits Th1 and Th2 development, and promotes Th17 differentiation from naive T cells. When used in combination with Retinoic acid, LY 294002 and CCG 1423, it induces intermediate mesoderm differentiation from ESCs. Inhibits growth of primary myeloma cells grown in the presence of bone marrow stromal cells. Cell-permeable.
Specifications
Specifications
| Synonym | JAK inhibitor I |
| Quantity | 10 mg |
| Molecular Formula | C18H16FN3O |
| Formula Weight | 309.34 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?