missing translation for 'onlineSavingsMsg'
Learn More

Pyrido[3,2-d][1,3]thiazole-2-thiol, 97%, Thermo Scientific™

Product Code. 10265644
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265644 5 g Amber Glass Bottle 5g
10100652 250 mg Amber Glass Bottle 250mg
10531322 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265644 Supplier Thermo Scientific Maybridge Supplier No. RH00329DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 57135-09-8
Molecular Formula C6H4N2S2
Molecular Weight (g/mol) 168.232
MDL Number MFCD00178760
InChI Key WITYIAHCBUYCSY-UHFFFAOYSA-N
Synonym thiazolo 5,4-b pyridine-2 1h-thione, thiazolo 5,4-b pyridine-2-thiol, 1,3 thiazolo 5,4-b pyridine-2-thiol, pyrido 3,2-d 1,3 thiazole-2-thiol, 2-mercaptothiazolo 5,4-b pyridine, pyrido 3,2-d-1,3-thiazol-2-thiol, 2-mercapto-1,3 thiazolo 5,4-b pyridine, 2-sulphanyl-1,3 thiazolo 5,4-b pyridine, 1h-1,3 thiazolo 5,4-b pyridine-2-thione, thiazolo 5,4-b pyridine-2-thione
PubChem CID 2782239
IUPAC Name 1H-[1,3]thiazolo[5,4-b]pyridine-2-thione
SMILES C1=CC2=C(N=C1)SC(=S)N2

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 168.24
Percent Purity 95%
Chemical Name or Material Pyrido[3, 2-d][1, 3]thiazole-2-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.