missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium p-Toluenesulfonate, TRC

Product Code. 30410962
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30410962 25 g
30418141 5 g
30425489 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30410962 Supplier Toronto Research Chemicals Supplier No. TRCP99160525G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24057-28-1
Molecular Formula C12H13NO3S
Molecular Weight (g/mol) 250.3
Synonym Pyridinium p-tosylate, Pyridinium Tosylate, 4-Methylbenzenesulfonic Acid compd. with Pyridine (1:1), Pyridine, 4-methylbenzenesulfonate, Pyridine, p-Toluenesulfonate, 4-Methylbenzenesulfonic Acid Pyridinium Salt, PPTS, Pyridinium 4-Toluenesulfonate, Pyridinium p-Toluenesulfonate
IUPAC Name 4-methylbenzenesulfonic acid;pyridine
SMILES Cc1ccc(cc1)S(=O)(=O)O.c2ccncc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C7H8O3S.C5H5N/c1-6-2-4-7(5-3-6)11(8,9)10;1-2-4-6-5-3-1/h2-5H,1H3,(H,8,9,10);1-5H
Formula Weight 251.0616
Chemical Name or Material Pyridinium p-Toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.