missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium dichromate, 98%

Product Code. 11388156
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388156 100 g 100g
11378156 25 g 25g
11398156 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388156 Supplier Thermo Scientific Alfa Aesar Supplier No. L15132.22
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alfa Aesar Pyridinium dichromate, 98%, Quantity: 100g, Melting Point: ∼145 deg.C to 147 deg.C, Molecular Weight: 376.205, Percent Purity: 98%, CAS: 20039-37-6, DOT Information: Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S. | CAS: 20039-37-6 | C10H12Cr2N2O7 | 376.205 g/mol

Pyridinium dichromate acts as a strong oxidizing agent used in the conversion of primary alcohols and secondary alcohols to aldehydes and ketones respectively. It plays an important role in the oxidation of unsaturated tertiary alcohols, silyl ethers, the carbon-boron bond, and oximes. Further, it is used in the conversion of acetals to esters and didehydroketones to enones in the presence of tert-butyl hydroperoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridinium dichromate acts as a strong oxidizing agent used in the conversion of primary alcohols and secondary alcohols to aldehydes and ketones respectively. It plays an important role in the oxidation of unsaturated tertiary alcohols, silyl ethers, the carbon-boron bond, and oximes. Further, it is used in the conversion of acetals to esters and didehydroketones to enones in the presence of tert-butyl hydroperoxide.

Solubility
Soluble in water, dimethyl formamide, dimethyl sulfoxide and acetonitrile. Slightly soluble in dichloromethane, chloroform and acetone. Insoluble in hexane, ether and ethyl acetate.

Notes
Hygroscopic. Incompatible withorganic materials and powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20039-37-6
Molecular Formula C10H12Cr2N2O7
Molecular Weight (g/mol) 376.205
MDL Number MFCD00013105
InChI Key LMYWWPCAXXPJFF-UHFFFAOYSA-P
Synonym pyridinium dichromate, cornforth reagent, pyridinium dichromate pdc, oxido-oxido dioxo chromio oxy-dioxochromium; pyridin-1-ium, pyridiniumdichromate, pyridinium-dichromate, dipyridinium dichromate, bis pyridium chromate, pyridiniumdichromate pdc, acmc-209f5g
PubChem CID 2724130
IUPAC Name oxido-(oxido(dioxo)chromio)oxy-dioxochromium;pyridin-1-ium
SMILES C1=CC=[NH+]C=C1.C1=CC=[NH+]C=C1.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Melting Point ∼145°C to 147°C
Quantity 100 g
UN Number UN3087
Sensitivity Hygroscopic
Solubility Information Soluble in water,dimethyl formamide,dimethyl sulfoxide and acetonitrile. Slightly soluble in dichloromethane,chloroform and acetone. Insoluble in hexane,ether and ethyl acetate.
Formula Weight 376.21
Percent Purity 98%
Chemical Name or Material Pyridinium dichromate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.