Learn More
Pyridinium dichromate, 98%
Beskrivelse
Pyridinium dichromate acts as a strong oxidizing agent used in the conversion of primary alcohols and secondary alcohols to aldehydes and ketones respectively. It plays an important role in the oxidation of unsaturated tertiary alcohols, silyl ethers, the carbon-boron bond, and oximes. Further, it is used in the conversion of acetals to esters and didehydroketones to enones in the presence of tert-butyl hydroperoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pyridinium dichromate acts as a strong oxidizing agent used in the conversion of primary alcohols and secondary alcohols to aldehydes and ketones respectively. It plays an important role in the oxidation of unsaturated tertiary alcohols, silyl ethers, the carbon-boron bond, and oximes. Further, it is used in the conversion of acetals to esters and didehydroketones to enones in the presence of tert-butyl hydroperoxide.
Solubility
Soluble in water, dimethyl formamide, dimethyl sulfoxide and acetonitrile. Slightly soluble in dichloromethane, chloroform and acetone. Insoluble in hexane, ether and ethyl acetate.
Notes
Hygroscopic. Incompatible withorganic materials and powdered metals.
Tekniske data
Tekniske data
| Melting Point | ∼145°C to 147°C |
| Quantity | 25 g |
| UN Number | UN3087 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water,dimethyl formamide,dimethyl sulfoxide and acetonitrile. Slightly soluble in dichloromethane,chloroform and acetone. Insoluble in hexane,ether and ethyl acetate. |
| Formula Weight | 376.21 |
| Percent Purity | 98% |
| Chemical Name or Material | Pyridinium dichromate |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.