missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium chlorochromate, 98%

Product Code. 11419303
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11419303

Brand: Thermo Scientific Alfa Aesar A11752.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26299-14-9 | C5H6ClCrNO3

Pyridinium chlorochromate is used as an oxidizing agent to convert primary and secondary alcohols to aldehydes and ketones respectively. It is involved in the preparation of cyclohexanone, (-)-pulegone and lactones. It plays an important role in the selective mono-oxidation of xylenes to tolualdehydes and arylhydroxyamines to nitroso compounds. Furthermore, it is utilized as an oxidant for amino acids, L-cystine, aniline, cycloalkanols, vicinal and non-vicinal diols as well as in the Babler oxidation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridinium chlorochromate is used as an oxidizing agent to convert primary and secondary alcohols to aldehydes and ketones respectively. It is involved in the preparation of cyclohexanone, (-)-pulegone and lactones. It plays an important role in the selective mono-oxidation of xylenes to tolualdehydes and arylhydroxyamines to nitroso compounds. Furthermore, it is utilized as an oxidant for amino acids, L-cystine, aniline, cycloalkanols, vicinal and non-vicinal diols as well as in the Babler oxidation reaction.

Solubility
Soluble in acetone, benzene, dichloromethane, acetonitrile and tetrahydrofuran.

Notes
Incompatible with alcohols, acetone and methanol.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26299-14-9
Molecular Formula C5H6ClCrNO3
MDL Number MFCD00013106
Synonym PCC

Specifications

Melting Point ∼206°C (decomposition)
Quantity 100 g
UN Number UN1479
Sensitivity Moisture sensitive
Merck Index 14,7974
Solubility Information Soluble in acetone,benzene,dichloromethane,acetonitrile and tetrahydrofuran.
Formula Weight 215.56
Percent Purity 98%
Chemical Name or Material Pyridinium chlorochromate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pyridinium chlorochromate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Oxidising solid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H317-May cause an allergic skin reaction.
  • H350i-May cause cancer by inhalation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Pyridinium chlorochromate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.