missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium chlorochromate, 98%

Product Code. 11429303
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429303 500 g 500g
11483990 25 g 25g
11419303 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429303 Supplier Thermo Scientific Alfa Aesar Supplier No. A11752.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26299-14-9 | C5H6ClCrNO3

Pyridinium chlorochromate is used as an oxidizing agent to convert primary and secondary alcohols to aldehydes and ketones respectively. It is involved in the preparation of cyclohexanone, (-)-pulegone and lactones. It plays an important role in the selective mono-oxidation of xylenes to tolualdehydes and arylhydroxyamines to nitroso compounds. Furthermore, it is utilized as an oxidant for amino acids, L-cystine, aniline, cycloalkanols, vicinal and non-vicinal diols as well as in the Babler oxidation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridinium chlorochromate is used as an oxidizing agent to convert primary and secondary alcohols to aldehydes and ketones respectively. It is involved in the preparation of cyclohexanone, (-)-pulegone and lactones. It plays an important role in the selective mono-oxidation of xylenes to tolualdehydes and arylhydroxyamines to nitroso compounds. Furthermore, it is utilized as an oxidant for amino acids, L-cystine, aniline, cycloalkanols, vicinal and non-vicinal diols as well as in the Babler oxidation reaction.

Solubility
Soluble in acetone, benzene, dichloromethane, acetonitrile and tetrahydrofuran.

Notes
Incompatible with alcohols, acetone and methanol.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26299-14-9
Molecular Formula C5H6ClCrNO3
MDL Number MFCD00013106
Synonym PCC

Specifications

Melting Point ∼206°C (decomposition)
Quantity 500 g
UN Number UN1479
Sensitivity Moisture sensitive
Merck Index 14,7974
Solubility Information Soluble in acetone,benzene,dichloromethane,acetonitrile and tetrahydrofuran.
Formula Weight 215.56
Percent Purity 98%
Chemical Name or Material Pyridinium chlorochromate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.