Learn More
Pyridinium chlorochromate, 98%, Thermo Scientific Chemicals
Description
Pyridinium chlorochromate is used as an oxidizing agent to convert primary and secondary alcohols to aldehydes and ketones respectively. It is involved in the preparation of cyclohexanone, (-)-pulegone and lactones. It plays an important role in the selective mono-oxidation of xylenes to tolualdehydes and arylhydroxyamines to nitroso compounds. Furthermore, it is utilized as an oxidant for amino acids, L-cystine, aniline, cycloalkanols, vicinal and non-vicinal diols as well as in the Babler oxidation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pyridinium chlorochromate is used as an oxidizing agent to convert primary and secondary alcohols to aldehydes and ketones respectively. It is involved in the preparation of cyclohexanone, (-)-pulegone and lactones. It plays an important role in the selective mono-oxidation of xylenes to tolualdehydes and arylhydroxyamines to nitroso compounds. Furthermore, it is utilized as an oxidant for amino acids, L-cystine, aniline, cycloalkanols, vicinal and non-vicinal diols as well as in the Babler oxidation reaction.
Solubility
Soluble in acetone, benzene, dichloromethane, acetonitrile and tetrahydrofuran.
Notes
Incompatible with alcohols, acetone and methanol.
Specifications
Melting Point | ∼206°C (decomposition) |
Quantity | 25 g |
UN Number | UN1479 |
Sensitivity | Moisture sensitive |
Merck Index | 14,7974 |
Solubility Information | Soluble in acetone,benzene,dichloromethane,acetonitrile and tetrahydrofuran. |
Formula Weight | 215.56 |
Percent Purity | 98% |
Chemical Name or Material | Pyridinium chlorochromate |
RUO – Research Use Only