missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, Ultrapure, Spectrophotometric Grade, 99.5+%

Product Code. 11338367
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
250mL
4 L
This item is not returnable. View return policy

Product Code. 11338367

Brand: Thermo Scientific Alfa Aesar 032436.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-86-1 | C5H5N | 79.102 g/mol

Pyridine is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point °C to 42°C
Color Colorless
Appearance Colorless liquid
Density 0.978 g/cm3 at 20°C
Boiling Point 115°C
Quantity 1 L
Flash Point 20°C (68°F)
Odor Unpleasant
Physical Form Liquid
Refractive Index 1.51
Assay Percent Range >99.5%
Vapor Pressure 20 hPa (15mm Hg) at 20°C
Beilstein 103233
Solubility Information Miscible with water,alcohol,ether,pet ether,and oils.
Formula Weight 79.1
Grade Spectrophotometric
Chemical Name or Material Pyridine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.