missing translation for 'onlineSavingsMsg'
Learn More

Pyridine (Sequencing), Fisher BioReagents™

Product Code. 10123053
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10123053 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10123053 Supplier Fisher BioReagents Supplier No. BP1155500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Pyridine is commonly used in automated synthesis protocols.
TRUSTED_SUSTAINABILITY

Specifications

pH 8.5
Melting Point -42°C
CAS 110-86-1
Color Colorless
Density 0.978g/cm³
Quantity 500 mL
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range ≥99.5 %
Molecular Formula C5H5N
Vapor Pressure 20mbar at 20°C
Viscosity 0.95 mPa/s at 20°C
UN Number 1282
Water 0.1% max.
Merck Index 15, 8081
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
Boiling Point 115°C
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES C1=CC=NC=C1
IUPAC Name pyridine
Molecular Weight (g/mol) 79.102
PubChem CID 1049
ChEBI CHEBI:16227
Formula Weight 79.1
Percent Purity ≥99.5%
Grade Sequencing
Chemical Name or Material Pyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.