missing translation for 'onlineSavingsMsg'
Learn More

Pyridine N-oxide, 95%

Product Code. 11458623
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458623 500 g 500g
11438623 25 g 25g
11448623 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458623 Supplier Thermo Scientific Alfa Aesar Supplier No. A10419.36
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 694-59-7 | C5H5NO | 95.101 g/mol

Pyridine N-oxide is used in the treatment of Kayser's disease. It is widely used in chemical industry. It can act as catalytic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Hygroscopic. Storage under a nitrogen blanket has been recommended. Store under 2-8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-59-7
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.101
MDL Number MFCD00006194
InChI Key ILVXOBCQQYKLDS-UHFFFAOYSA-N
Synonym pyridine-n-oxide, pyridine 1-oxide, pyridine n-oxide, pyridine oxide, pyridine, 1-oxide, pyridine-1-oxide, unii-91f12jjj4h, py n-oxide, pyridin-1-ium-1-olate, pyridine-oxide
PubChem CID 12753
ChEBI CHEBI:29136
IUPAC Name 1-oxidopyridin-1-ium
SMILES C1=CC=[N+](C=C1)[O-]

Specifications

Melting Point 61°C to 66°C
Boiling Point 270°C
Flash Point 143°C (289°F)
Quantity 500 g
Beilstein 105257
Sensitivity Very Hygroscopic
Merck Index 14,7972
Solubility Information Soluble in water.
Formula Weight 95.1
Percent Purity 95%
Chemical Name or Material Pyridine N-oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.