missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrochloride, 98%

Product Code. p-7081061
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11404223

Brand: Thermo Scientific Alfa Aesar A10871.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 628-13-7 | C5H6ClN | 115.56 g/mol

Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

Solubility
Soluble in water (85 g/100 ml) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-13-7
Molecular Formula C5H6ClN
Molecular Weight (g/mol) 115.56
MDL Number MFCD00012802
InChI Key AOJFQRQNPXYVLM-UHFFFAOYSA-N
Synonym pyridine hydrochloride, pyridine, hydrochloride, pyridinehydrochloride, pyridine hcl, pyridine, hydrochloride 1:1, pyridine-hcl, pyridine.hcl, pyridiniumchloride, pyridine hcl salt, pyrdine hydrochloride
PubChem CID 10176127
IUPAC Name pyridine;hydrochloride
SMILES C1=CC=NC=C1.Cl

Specifications

Melting Point ∼145°C
Boiling Point 222°C to 224°C
Quantity 50 g
Beilstein 3615340
Sensitivity Hygroscopic
Solubility Information Soluble in water (85g/100 ml) and methanol.
Formula Weight 115.56
Percent Purity 98%
Chemical Name or Material Pyridine hydrochloride
compliance-icons
Product Identifier
  • Pyridine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Pyridine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.