missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrochloride, 98%

Product Code. 11414223
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414223 250 g 250g
11404223 50 g 50g
11494213 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414223 Supplier Thermo Scientific Alfa Aesar Supplier No. A10871.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 628-13-7 | C5H6ClN | 115.56 g/mol

Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

Solubility
Soluble in water (85 g/100 ml) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-13-7
Molecular Formula C5H6ClN
Molecular Weight (g/mol) 115.56
MDL Number MFCD00012802
InChI Key AOJFQRQNPXYVLM-UHFFFAOYSA-N
Synonym pyridine hydrochloride, pyridine, hydrochloride, pyridinehydrochloride, pyridine hcl, pyridine, hydrochloride 1:1, pyridine-hcl, pyridine.hcl, pyridiniumchloride, pyridine hcl salt, pyrdine hydrochloride
PubChem CID 10176127
IUPAC Name pyridine;hydrochloride
SMILES C1=CC=NC=C1.Cl

Specifications

Melting Point ∼145°C
Boiling Point 222°C to 224°C
Quantity 250 g
Beilstein 3615340
Sensitivity Hygroscopic
Solubility Information Soluble in water (85g/100 ml) and methanol.
Formula Weight 115.56
Percent Purity 98%
Chemical Name or Material Pyridine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.