missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, Solstice

Product Code. 16675743
Change view
Click to view available options
Quantity:
35 kg
Unit Size:
35kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16675743 35 kg 35kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16675743 Supplier Fluka Supplier No. 16037S35KG
  • ONLINE EXCLUSIVE PRICE
    £1060.00
    £887.55 / 35kg
    Save £172.45
    16% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 110-86-1
CAS Max % 1
Melting Point -42°C
Color Colorless
Density 0.98 g/cm3 (20°C)
pH 8.8
Boiling Point 115°C (1013 hPa)
Flash Point 20°C
Molecular Formula C5H5N
Quantity 35 kg
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES C1=CC=NC=C1
IUPAC Name pyridine
Molecular Weight (g/mol) 79.10
Chemical Name or Material Pyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.