Learn More
Pyridine-d5, for NMR, with 0.03% TMS, 100.0 atom% D, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7291-22-7, 75-76-3 |
---|---|
Molecular Formula | C5H5N |
Molecular Weight (g/mol) | 84.13 |
MDL Number | MFCD00044639 |
InChI Key | JUJWROOIHBZHMG-RALIUCGRSA-N |
Synonym | pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c |
PubChem CID | 558519 |
IUPAC Name | 2,3,4,5,6-pentadeuteriopyridine |
SMILES | [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10326641
|
Thermo Scientific Acros
351490050 |
5 mL | Glass Bottle |
N/A
|
N/A | ||||
10336641
|
Thermo Scientific Acros
351490100 |
10 mL | Glass Bottle |
N/A
|
N/A | ||||
10193521
|
Thermo Scientific Acros
351490250 |
25 mL | Glass Bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
7291-22-7, 75-76-3 | |
84.13 | |
JUJWROOIHBZHMG-RALIUCGRSA-N | |
558519 | |
[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H] |
C5H5N | |
MFCD00044639 | |
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c | |
2,3,4,5,6-pentadeuteriopyridine |
Specifications
Liquid | |
1.0500g/mL | |
20°C | |
0.02% max. (Coulometric) | |
-41.0°C | |
C5H5N | |
20, III, 2305 | |
JUJWROOIHBZHMG-RALIUCGRSA-N | |
2,3,4,5,6-pentadeuteriopyridine | |
558519 | |
NMR |
Colorless | |
99.95% min. | |
1.507 | |
7291-22-7, 75-76-3 | |
114.4°C | |
MFCD00044639 | |
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c | |
[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H] | |
84.13 | |
84.14 | |
Pyridine-d5, 100.0 atom% D |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 230-720-2
- Pyridine-d5
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only