missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-d{5}, 99.5% (Isotopic), Thermo Scientific Chemicals

Product Code. 11387928
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Pyridine-d{5} is used in the synthesis complexes with cobalt(II) halides, pyridine-d5, a deuterated form of pyridine, ionic liquid matrices (ILMs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7291-22-7
Molecular Formula C5H5N
Molecular Weight (g/mol) 84.13
MDL Number MFCD00044639
InChI Key JUJWROOIHBZHMG-RALIUCGRSA-N
Synonym pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
PubChem CID 558519
IUPAC Name 2,3,4,5,6-pentadeuteriopyridine
SMILES [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]

Specifications

Physical Form Liquid
Density 1.05 g/mL
Flash Point 20°C (68°F)
Odor Unpleasant
Refractive Index 1.506
Quantity 2 g
Melting Point -42°C
Boiling Point 115°C
UN Number UN1282
Beilstein 114377
Solubility Information Soluble in water,alcohol,ether,acetone,and benzene.
Formula Weight 84.14
Percent Purity 99.5%
Chemical Name or Material Pyridine-d5, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'