missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-d{5}, 100%(Isotopic)

Product Code. 11344438
Change view
Click to view available options
Quantity:
10 Ea.
2 Ea.
Unit Size:
Pack of 10
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344438 10 Ea. Pack of 10
11354438 2 Ea. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11344438 Supplier Thermo Scientific Alfa Aesar Supplier No. 042343.KV
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7291-22-7 | C5H5N | 84.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7291-22-7
Molecular Formula C5H5N
Molecular Weight (g/mol) 84.13
MDL Number MFCD00044639
InChI Key JUJWROOIHBZHMG-RALIUCGRSA-N
Synonym pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
PubChem CID 558519
IUPAC Name 2,3,4,5,6-pentadeuteriopyridine
SMILES [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]

Specifications

Physical Form Liquid
Density 1.05g/mL
Flash Point 20°C (68°F)
Odor Unpleasant
Packaging Prescored Ampule
Refractive Index 1.506
Quantity 10 Ea.
Melting Point -42°C
Boiling Point 115°C
UN Number UN1282
Beilstein 114377
Formula Weight 84.14
Percent Purity 100%
Chemical Name or Material Pyridine-d5, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.