Learn More
Pyridine, 99+%, for analysis, Thermo Scientific Chemicals
£20.00 - £455.00
Chemical Identifiers
CAS | 110-86-1 |
---|---|
Molecular Formula | C5H5N |
Molecular Weight (g/mol) | 79.1 |
MDL Number | MFCD00011732 |
InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
Synonym | azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 |
PubChem CID | 1049 |
ChEBI | CHEBI:16227 |
IUPAC Name | pyridine |
SMILES | C1=CC=NC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10341054
|
Thermo Scientific Acros
290861000 |
100 mL | Glass bottle |
£20.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10200214
|
Thermo Scientific Acros
290860010 |
1 L | Glass bottle |
£74.50
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10637684
|
Thermo Scientific Acros
290860025 |
2.5 L | Glass bottle |
£154.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11367030
|
Thermo Scientific Acros
290860100 |
10 L | Plastic drum |
£455.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-86-1 | |
79.1 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
1049 | |
pyridine |
C5H5N | |
MFCD00011732 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
CHEBI:16227 | |
C1=CC=NC=C1 |
Specifications
110-86-1 | |
0.9780g/mL | |
17°C | |
C5H5N | |
MFCD00011732 | |
01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153 | |
0.978 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents | |
C1=CC=NC=C1 | |
79.1 | |
CHEBI:16227 | |
79.1 | |
Analysis | |
Pyridine |
-42.0°C | |
115.0°C to 116.0°C | |
99% min. (GC) | |
1.5085 to 1.5105 | |
20,181 | |
15,8081 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
1049 | |
0.95 mPa.s (20°C) | |
99+% | |
Liquid |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-809-9
RUO – Research Use Only