missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, >99%, MP Biomedicals™

Product Code. 11464231 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464231 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11464231 Supplier MP Biomedicals Supplier No. 0215198383
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pyridine acts as a solvent for anhydrous mineral salts, organic and inorganic compounds.

  • Pyridine is a synthetic intermediate in laboratory and industry.
  • Its weakly basic nature allows it to act as a proton acceptor in acid base reactions, and to form salts with strong acids.
  • In the acylation of alcohols, pyridine can act as a nucleophilic catalyst.
  • Various reports have described the use of pyridine to prepare such compounds as indanones, 2 cyclopentenones, polycarbonates, symmetric aromatic trisulfides and tetrasulfides, and calcium-η 2-pyrazolato complexes.

Specifications

CAS 110-86-1
Color Colorless to Yellow
Density 0.982 (water = 1)
Assay Percent Range >99%
Molecular Formula C5H5N
Quantity 250 mL
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES C1=CC=NC=C1
IUPAC Name pyridine
Molecular Weight (g/mol) 79.102
PubChem CID 1049
ChEBI CHEBI:16227
Formula Weight 79.1
Physical Form Liquid
Chemical Name or Material Pyridine
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.