missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-carboxaldehyde, 97%

Product Code. 11443404
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443404 25 g 25g
11453404 100 g 100g
11463404 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11443404 Supplier Thermo Scientific Alfa Aesar Supplier No. A10656.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 872-85-5 | C6H5NO | 107.112 g/mol

Pyridine-4-carboxaldehyde is an heterocyclic building block used for the synthesis of various pharmaceutical compounds, such as new 1,4-dihydropyridin-4-yl-phenoxyacetohydrazones, having anticonvulsant and anti-inflammatory properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-4-carboxaldehyde is an heterocyclic building block used for the synthesis of various pharmaceutical compounds, such as new 1,4-dihydropyridin-4-yl-phenoxyacetohydrazones, having anticonvulsant and anti-inflammatory properties.

Solubility
Soluble in water (20 mg/ml at 20°C), ethanol and acetone.

Notes
It is air and light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-85-5
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.112
MDL Number MFCD00006425
InChI Key BGUWFUQJCDRPTL-UHFFFAOYSA-N
Synonym 4-pyridinecarboxaldehyde, isonicotinaldehyde, 4-formylpyridine, isonicotinic aldehyde, p-pyridinealdehyde, pyridine-4-carboxaldehyde, 4-pyridinealdehyde, 4-pyridylaldehyde, 4-pyridinecarbaldehyde, pyridine-4-aldehyde
PubChem CID 13389
IUPAC Name pyridine-4-carbaldehyde
SMILES C1=CN=CC=C1C=O

Specifications

Melting Point -4°C
Density 1.132
Boiling Point 198°C
Flash Point 75°C (167°F)
Refractive Index 1.544
Quantity 25 g
Beilstein 105342
Sensitivity Air and light sensitive
Solubility Information Soluble in water (20mg/ml at 20°C),ethanol and acetone.
Formula Weight 107.11
Percent Purity 97%
Chemical Name or Material Pyridine-4-carboxaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.