missing translation for 'onlineSavingsMsg'
Learn More

pyridine-3-sulfonamide, TRC

Product Code. 30389732
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389732 10 mg
30406858 50 mg
30394760 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389732 Supplier Toronto Research Chemicals Supplier No. TRCI70695310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2922-45-4
Molecular Formula C5H6N2O2S
Molecular Weight (g/mol) 158.18
IUPAC Name pyridine-3-sulfonamide
SMILES NS(=O)(=O)c1cccnc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C5H6N2O2S/c6-10(8,9)5-2-1-3-7-4-5/h1-4H,(H2,6,8,9)
Formula Weight 158.015
Chemical Name or Material pyridine-3-sulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.