missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-carboxamidine hydrochloride 0.5 hydrate, 95%, Thermo Scientific™

Product Code. 10091044
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10091044 25 g 25g
10118164 5 g 5g
10453373 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10091044 Supplier Thermo Scientific Maybridge Supplier No. MO07766EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 871825-82-0
Molecular Formula C12H18Cl2N6O
Molecular Weight (g/mol) 333.217
InChI Key AOHLMHDFGXVZAQ-UHFFFAOYSA-N
Synonym pyridine-3-carboximidamide hemihydrate hydrochloride
PubChem CID 90477204
IUPAC Name pyridine-3-carboximidamide;hydrate;dihydrochloride
SMILES C1=CC(=CN=C1)C(=N)N.C1=CC(=CN=C1)C(=N)N.O.Cl.Cl

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 25 g
Formula Weight 166.61
Percent Purity 95%
Chemical Name or Material Pyridine-3-carboxamidine hydrochloride 0.5 hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.