missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-boronic acid

Product Code. 11348126
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348126 5 g 5g
11338126 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348126 Supplier Thermo Scientific Alfa Aesar Supplier No. L15040.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1692-25-7 | C5H6BNO2 | 122.92 g/mol

Reactant used for preparation of phosphine-free Suzuki-Miyaura cross-coupling reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intermolecular aminocarbonylation and annulation, N-arylation using copper acetylacetonate catalys, copper-mediated cyanation and regioselective cyanation of electron-rich benzenes, New linear poly(phenylpyridyl) chains by Suzuki coupling and Oligopyridyl foldamers as mimics of a-helix twist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for preparation of phosphine-free Suzuki-Miyaura cross-coupling reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intermolecular aminocarbonylation and annulation, N-arylation using copper acetylacetonate catalys, copper-mediated cyanation and regioselective cyanation of electron-rich benzenes, New linear poly(phenylpyridyl) chains by Suzuki coupling and Oligopyridyl foldamers as mimics of a-helix twist.

Solubility
Slightly soluble in methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1692-25-7
Molecular Formula C5H6BNO2
Molecular Weight (g/mol) 122.92
MDL Number MFCD00674177
InChI Key ABMYEXAYWZJVOV-UHFFFAOYSA-N
Synonym pyridine-3-boronic acid, 3-pyridylboronic acid, 3-pyridineboronic acid, pyridin-3-yl boronic acid, 3-pyridinylboronic acid, pyridin-3-yl-3-boronic acid, 3-pyridineboric acid, 3-pyridinebornic acid, pyridyl-3-boronic acid, pyridine 3-boronic acid
PubChem CID 2734378
IUPAC Name pyridin-3-ylboronic acid
SMILES OB(O)C1=CC=CN=C1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 471943
Solubility Information Slightly soluble in methanol.
Formula Weight 122.92
Chemical Name or Material Pyridine-3-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.