missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3,4-dicarboxylic anhydride, 97%

Product Code. 11469672
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469672 1 g 1g
11479672 5 g 5g
11489672 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469672 Supplier Thermo Scientific Alfa Aesar Supplier No. A10771.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4664-08-8 | C7H3NO3 | 149.11 g/mol

Pyridine-3,4-dicarboxylic anhydride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-3,4-dicarboxylic anhydride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Decomposes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4664-08-8
Molecular Formula C7H3NO3
Molecular Weight (g/mol) 149.11
MDL Number MFCD00010680
InChI Key KFKMGUPDWTWQFM-UHFFFAOYSA-N
Synonym furo 3,4-c pyridine-1,3-dione, 3,4-pyridinedicarboxylic anhydride, pyridine-3,4-dicarboxylic anhydride, 3,4-pyridinedicarboxylic acid anhydride, 1h,3h-furo 3,4-c pyridine-1,3-dione, cinchomeronic anhydride, pyridine-3,4-dicarboxylicanhydride, 1,3-dihydrofuro 3,4-c pyridine-1,3-dione, furano 3,4-c pyridine-1,3-dione, 3,4-pyridinecarboxylic acid anhydride
PubChem CID 278398
IUPAC Name furo[3,4-c]pyridine-1,3-dione
SMILES C1=CN=CC2=C1C(=O)OC2=O

Specifications

Melting Point 75°C to 79°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 149.11
Percent Purity 97%
Chemical Name or Material Pyridine-3,4-dicarboxylic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.