missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-carboxamide (Picolinamide), Mikromol™

Product Code. 17922686
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17922686 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17922686 Supplier Mikromol Supplier No. MM0552.030025
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pyridine-2-carboxamide (Picolinamide)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Picolinamide
CAS 1452-77-3
Analyte or Component Names Picolinamide
Shipping Condition Room Temperature
Molecular Formula C6 H6 N2 O
Certifications/Compliance ISO 17025
Synonym 2-Pyridinecarboxamide, Picolinamide (6CI,7CI,8CI), 2-Carbamoylpyridine, NSC 524473, Picolinic acid amide, Picolinoylamide, Pyridine-2-carboxamide, α-(Aminocarbonyl)pyridine, α-Picolinamide, α-Picolinic acid amide, α-Pyridinecarboxamide, Nicotinamide Impurity C
InChI Formula InChI=1S/C6H6N2O/c7-6(9)5-3-1-2-4-8-5/h1-4H,(H2,7,9)
SMILES NC(=O)c1ccccn1
IUPAC Name pyridine-2-carboxamide
Molecular Weight (g/mol) 122.12
Quantity 25 mg
Formula Weight 122.048 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.