missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-carboxaldehyde, 99%

Product Code. 11480213
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480213 100 g 100g
11414000 25 g 25g
11490213 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480213 Supplier Thermo Scientific Alfa Aesar Supplier No. A11391.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1121-60-4 | C6H5NO | 107.112 g/mol

Pyridine-2-carboxaldehyde, 99% is used as an Intermediate of Bisacodyl. It is also used as an intermediate in pharamceutical research, Intermediate in organicc synethesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-2-carboxaldehyde, 99% is used as an Intermediate of Bisacodyl. It is also used as an intermediate in pharamceutical research, Intermediate in organicc synethesis.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C .Store under inert gas. Stable under recommended storage conditions.Incompatible to Strong bases, Strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-60-4
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.112
MDL Number MFCD00006290
InChI Key CSDSSGBPEUDDEE-UHFFFAOYSA-N
Synonym 2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde
PubChem CID 14273
ChEBI CHEBI:73012
IUPAC Name pyridine-2-carbaldehyde
SMILES C1=CC=NC(=C1)C=O

Specifications

Melting Point -21°C
Density 1.124
pH 6.5
Boiling Point 180°C to 181°C
Flash Point 68°C (154°F)
Odor Characteristic
Refractive Index 1.536
Quantity 100 g
UN Number UN2810
Beilstein 105341
Sensitivity Air and light sensitive
Solubility Information It is soluble in water.
Formula Weight 107.11
Percent Purity 99%
Chemical Name or Material Pyridine-2-carboxaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyridine-2-carboxaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Pyridinecarboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.