missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-boronic acid dimethyl ester, 95%, Thermo Scientific™

Product Code. 15449837
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449837 250 mg 250mg
15439837 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449837 Supplier Thermo Scientific Alfa Aesar Supplier No. H53067.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136805-54-4
Molecular Formula C7H10BNO2
Molecular Weight (g/mol) 150.972
MDL Number MFCD03412109
InChI Key ITSIDKYMJQVPQQ-UHFFFAOYSA-N
Synonym dimethyl pyridin-2-ylboronate, pyridine-2-boronic acid dimethyl ester, dimethyl pyridineboronate, pyridine-2-boronic acid, dimethyl ester, dimethoxy pyridin-2-yl borane, 2-pyridylboronic acid, dimethyl ester, 2-pyridineboronic acid dimethyl ester, akos brn-0425, 2-dimethoxyboryl pyridine, acmc-209ca1
PubChem CID 2763164
IUPAC Name dimethoxy(pyridin-2-yl)borane
SMILES B(C1=CC=CC=N1)(OC)OC

Specifications

Quantity 250 mg
Formula Weight 150.97
Percent Purity 95%
Chemical Name or Material Pyridine-2-boronic acid dimethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.