missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-boronic acid, 95%

Product Code. 15490058
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490058 1 g 1g
15400068 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490058 Supplier Thermo Scientific Alfa Aesar Supplier No. H53314.03
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 197958-29-5 | C5H6BNO2 | 122.92 g/mol

Pyridine-2-boronic acid is used as a key starting material for the formation of pyridyl boronic ester such as pyridine-2-boronic acid dimethyl ester. Furthermore, the dimethyl ester reacts with bromquinoline to prepare the 2-pyridyl derivative of quinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-2-boronic acid is used as a key starting material for the formation of pyridyl boronic ester such as pyridine-2-boronic acid dimethyl ester. Furthermore, the dimethyl ester reacts with bromquinoline to prepare the 2-pyridyl derivative of quinoline.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 197958-29-5
Molecular Formula C5H6BNO2
Molecular Weight (g/mol) 122.92
MDL Number MFCD00151844
InChI Key UMLDUMMLRZFROX-UHFFFAOYSA-N
Synonym 2-pyridineboronic acid, pyridine-2-boronic acid, 2-pyridinylboronic acid, 2-pyridylboronic acid, pyridin-2-boronic acid, pyridylboronic acid, boronic acid, 2-pyridinyl, 2-pyridineboronicacid, 2-pyridinyl-boronic acid, pyridinylboronic acid
PubChem CID 2762745
IUPAC Name pyridin-2-ylboronic acid
SMILES OB(O)C1=CC=CC=N1

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 122.92
Percent Purity 95%
Chemical Name or Material Pyridine-2-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.