missing translation for 'onlineSavingsMsg'
Learn More

Pyridin-2-amine (2-Pyridylamine), Mikromol™

Product Code. 18063250
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18063250 25 mg 25mg
18063440 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18063250 Supplier Mikromol Supplier No. MM0084.010025
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pyridin-2-amine (2-Pyridylamine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminopyridine
CAS 504-29-0
Analyte or Component Names 2-Aminopyridine
Shipping Condition Room Temperature
Molecular Formula C5 H6 N2
Certifications/Compliance ISO 17025
Synonym Pyridin-2-amine, Mepyramine Maleate Imp. C (EP), Tenoxicam Imp. A (EP), Piroxicam Imp. A (EP), 2-Pyridylamine, 2-Aminopyridine, Mepyramine Maleate Impurity C, Piroxicam Impurity A, Tenoxicam Impurity A
InChI Formula InChI=1S/C5H6N2/c6-5-3-1-2-4-7-5/h1-4H,(H2,6,7)
SMILES Nc1ccccn1
IUPAC Name pyridin-2-amine
Molecular Weight (g/mol) 94.11
Quantity 25 mg
Formula Weight 94.0531 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.