missing translation for 'onlineSavingsMsg'
Learn More

Pyridazine, 98+%

Product Code. 11440813
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440813 5 g 5g
11450813 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440813 Supplier Thermo Scientific Alfa Aesar Supplier No. A13266.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 289-80-5 | C4H4N2 | 80.09 g/mol

Pyrrolo[1,2-b]pyridazine 1 is a N-bridgehead aromatic heterocycle containing two nitrogen atoms, is obtained by the condensation of pyridazine and pyrrole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrolo[1,2-b]pyridazine 1 is a N-bridgehead aromatic heterocycle containing two nitrogen atoms, is obtained by the condensation of pyridazine and pyrrole.

Solubility
Miscible with water, benzene, DMF. Soluble in alcohol, ether

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 289-80-5
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006463
InChI Key PBMFSQRYOILNGV-UHFFFAOYSA-N
Synonym orthodiazine, 1,2-diazine, o-diazine, 1,2-diazabenzene, pyridazin, 1,2-diazin, oizine, pyridazine, pubchem17789, acmc-209h5x
PubChem CID 9259
ChEBI CHEBI:30954
IUPAC Name pyridazine
SMILES C1=CC=NN=C1

Specifications

Melting Point -8°C
Density 1.105
Boiling Point 87°C (14 mmHg)
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.524
Quantity 5 g
Beilstein 103906
Merck Index 14,7969
Solubility Information Miscible with water, benzene, DMF; Soluble in alcohol, ether
Formula Weight 80.09
Percent Purity ≥98%
Chemical Name or Material Pyridazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.