Learn More
Nickel Electro™ Clifton™ Stainless Steel Spoon One End Spatulas
Contains a spatula on one end and a spoon on the other. Nickel-Electro™ Stainless Steel Spoon One End Spatulas are quick to clean with an attractive shiny appearance making it the choice for applications requiring sterile surfaces.
Brand:  Nickel Electro™ 2012
Product Code. 11716617
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Contains a Laboratory Spoon on one end, and a spatula on the other.
  • Stainless steel, non-magnetic with bright finish
  • 180mm long
  • Spoon: 20mm wide
  • Spoon capacity: 2mL
  • Supplied in packs of 10