missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Screwcap Spare Inserts
For use with Black SVL™ PBT screwcaps
Brand: Pyrex 4513/18M
Code : A0
Additional Details : Weight : 0.01100kg
Description
- PTFE faced sealing disc for chemical resistance
- Autoclavable up to 121°C
Specifications
Screw cap | |
PTFE, Rubber | |
Autoclavable | |
18 mm |
20/Pk. | |
Spare insert | |
Spare Insert | |
PTFE faced sealing disc |