missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Plastic Coated Borosilicate Glass Wide Neck Round Media Bottles Without Cap

Product Code. 10308859
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10308859 10,000 mL Each
12539135 100 mL Pack of 10
11962629 250 mL Pack of 10
12069012 500 mL Pack of 10
12559135 1000 mL Pack of 10
10221560 2000 mL Pack of 10
10596340 5000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10308859 Supplier Pyrex Supplier No. 1518/18
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from Borosilicate glass with clear plastic coating to provide added protection against mechanical breakage. Pyrex™ Plastic Coated Borosilicate Glass Wide Neck Round Media Bottles Without Cap reduces the hazard of flying glass and liquid splashes in the event of breakage. Coating withstands steam sterilisation up to 135° C. Bottles are supplied without cap or pouring ring.

  • Bottles only. Supplied without cap or pouring ring
  • Borosilicate glass with clear plastic coating to provide added protection against mechanical breakage
  • Reduces the hazard of flying glass and liquid splashes in the event of breakage
  • Printed with retrace code: downloadable batch certificate
  • Coating withstands steam sterilisation up to 135°C
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10,000 mL
Coating Plastic
Color Clear
For Use With (Application) Ideal for mixing and storage of culture medium
Material Borosilicate Glass
Certifications/Compliance ISO 4796
Graduated Yes
Mouth Wide
Shape Round
Height (Metric) 410 mm
Closure Size 45 mm
Diameter (Metric) 227 mm
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.