missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Narrow-Mouth Reagent Bottle

Product Code. 11377874
Change view
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
15,000 mL
20,000 mL
2000 mL
250 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11377874 15,000 mL Each
12599845 250 mL Pack of 10
12539885 500 mL Pack of 10
12621992 1000 mL Pack of 10
11347874 2000 mL Pack of 10
11357874 5000 mL Each
11367874 10,000 mL Each
11387874 20,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 11377874 Supplier Pyrex Supplier No. 1526/20D
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from Pyrex™ superior strength borosilicate glass. Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles are ideal for storing chemicals in the laboratory. Use rubber stoppers (not included) to secure.

  • Unground neck for use with rubber stoppers (not supplied)
  • Borosilicate glass provides strong chemical resistance
  • Complies with ISO 4796
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 15,000 mL
Material Borosilicate Glass
Neck ID (Metric) 59mm
Quantity Each
Neck Finish Unground
Certifications/Compliance ISO 4796
Closure Included No
Mouth Narrow
Height (Metric) 460 mm
Diameter (Metric) 258 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.